missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Bottle Jacket

Provides added protection of a polyethylene bottle without contaminating high-purity solvents

Supplier:  Justrite™ JMC12009

Catalog No. 17-985-79


Only null left
Explore available promotions

Description

Description

  • Ideal for 4L and 1 gal. bottles
  • Place a bottle in the bottom half of the jacket and rotate lid for secure containment
  • Handle offers balanced pouring
  • Acts as a shock absorber to minimize breakage and spills due to bumps or falls
Specifications

Specifications

Bottle Jacket
Bottles (4 L, 1 gal.)
35.5 cm
14 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions