missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Justrite™ EcoPolyBlend™ Spill Control Pallets
Made of up to 100% recycled polyethylene
Supplier: Justrite™ 28628
Includes: Two and four drum square pallets include wide forklift pockets.
Description
- Protect against groundwater contamination, using post-industrial material
- Robust pallets withstand the toughest environments for superior spill control
- Durable, UV-protected polyethylene is nearly impervious to chemical attack from hazardous acids, caustics, and solvents
- Seamless construction eliminates leaks—even if a 55-gallon (208L) drum fails and completely empties
- Oversized leakproof sumps meet or exceed regulations
- Sturdy, removable 2.5 in. (64mm) thick grates offer superior crack resistance; self-locating
- Black pallets constructed of 100% recycled content; dark color camouflages dirt and keeps work places looking clean
- Yellow pallets ideal for high visibility or liquid segregation; virgin poly sumps with 100% recycled grates (a total of 35% recycled content)
- Square pallet may be equipped with an optional ramp
- Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
- Recycled: Item contains recycled content as described in the item detail
Compliance
Meet or exceed EPA 40 CFR 264.175; NFPA 1 the Uniform Fire Code 60.3.2.10; International Fire Code 2704.2.3; Spill Prevention, Control and Countermeasures (SPCC) Compliant; NFPA 30 9.13.1 and OSHA 29 CFR 1910.106(e)(2)(iii).
Specifications
SPCC, EPA, NFPA, U (NFPA), meets fire codes | |
75 gal. | |
Polyethylene | |
Keep workers safe from accidental slips indoors and avoid costly hazmat cleanup costs, protect against groundwater contamination | |
3 Drums | |
3750 lb. | |
Chemical | |
185.4 cm | |
29.5 cm | |
63.5 cm |
Yellow | |
284 L | |
Ecopolyblend Spill Control Pallet | |
Drums | |
3 drums with drain | |
1701 kg | |
73 in. | |
11.6 in. | |
25 in. |