missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Justrite™ EcoPolyBlend™ Spill Control Pallets

Made of up to 100% recycled polyethylene

Supplier:  Justrite™ 28631

 View more versions of this product

Catalog No. 19-159-660


Explore available promotions

Includes: Two and four drum square pallets include wide forklift pockets.

Description

Description

  • Protect against groundwater contamination, using post-industrial material
  • Robust pallets withstand the toughest environments for superior spill control
  • Durable, UV-protected polyethylene is nearly impervious to chemical attack from hazardous acids, caustics, and solvents
  • Seamless construction eliminates leaks—even if a 55-gallon (208L) drum fails and completely empties
  • Oversized leakproof sumps meet or exceed regulations
  • Sturdy, removable 2.5 in. (64mm) thick grates offer superior crack resistance; self-locating
  • Black pallets constructed of 100% recycled content; dark color camouflages dirt and keeps work places looking clean
  • Yellow pallets ideal for high visibility or liquid segregation; virgin poly sumps with 100% recycled grates (a total of 35% recycled content)
  • Square pallet may be equipped with an optional ramp
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail

Compliance

Meet or exceed EPA 40 CFR 264.175; NFPA 1 the Uniform Fire Code 60.3.2.10; International Fire Code 2704.2.3; Spill Prevention, Control and Countermeasures (SPCC) Compliant; NFPA 30 9.13.1 and OSHA 29 CFR 1910.106(e)(2)(iii).

Specifications

Specifications

SPCC, EPA, NFPA, U (NFPA), meets fire codes
75 gal.
Polyethylene
Keep workers safe from accidental slips indoors and avoid costly hazmat cleanup costs, protect against groundwater contamination
4 Drums
5000 lb.
Chemical
246.4 cm
22.9 cm
63.5 cm
Black
28.4 L
Ecopolyblend Spill Control Pallet
Drums
4 Drum In-Line
2268 kg
97 in.
9 in.
25 in.
Videos
SDS
Documents

Documents

Promotions

Promotions