missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Justrite™ Flammables Single-Door Benchtop Cabinet
Allows every lab to have a small flammables storage cabinet
Supplier: Justrite™ 890400
Includes: One adjustable shelf
Description
Ideal for localized storage of small quantities of flammable laboratory chemicals.
Features of Justrite Flammables Cabinets
- Double-wall construction provides 1.5 in. of insulating air space to help prevent the spread of flames
- High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
- Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
- Easy-closing, self-latching doors facilitate fire protection of the cabinet
- Self latch engages critical three-point latching system for protection under fire conditions
- SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
- Exterior side panel has built-in grounding connector for easy grounding
- Finished inside and out with durable lead-free powder paint
- 1.5 in. (3.8cm) air space between 18G steel walls
- 2 in. (5cm) vent openings
- With manual closing door
Order Info
Capacity: 4 gal. (15L). Dimensions: 17L x 17W x 22 in.H (43 x 43 x 56cm). Weight: 62 lb. (28kg).
Compliance
Meet OSHA and NFPA requirements
Warranty
10-Year
Specifications
34.9 cm | |
22 in. | |
Steel (CR) | |
28.12 kg | |
13.75 in. | |
43.2 cm | |
4 gal. | |
13.75 in. | |
47 cm | |
Flammables | |
43.2 cm | |
55.9 cm | |
1 |
34.9 cm | |
Epoxy, Polyester | |
One adjustable shelf, U-Loc™ handle with a cylinder lock and key set or an optional padlock, Haz-Alert™ reflective trilingual warning labels, leakproof 2 in. bottom sump, dual vents with flame arresters, four adjustabl | |
17 in. | |
13.75 x 13.75 x 18.5 in. (34.9 x 34.9 x 47 cm) | |
62 lb. | |
1 | |
18.5 in. | |
Countertop Safety Cabinet | |
17 in. | |
Yellow | |
Manual-close | |
NFPA, OSHA |