missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Justrite™ Sure-Grip™ EX Slimline Flammable Safety Cabinet
Add storage in tight spaces by adding a Justrite Sure-Grip™ EX Slimline Flammable Safety Cabinet to your lab.
$2,316.41 - $2,821.71
Specifications
Depth (English) Interior | 14.56 in. |
---|---|
Depth (Metric) Exterior | 45.7 cm |
Depth (Metric) Interior | 37 cm |
Color | Yellow |
Depth (English) Exterior | 18 in. |
Catalog Number | Mfr. No. | Door Style | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Door Style | Price | Quantity | |||||
19160387
|
Justrite™
892220 |
Self-close |
N/A
|
|
|||||
19-160-377
|
Justrite™
892200 |
Manual-close |
N/A
|
|
|||||
Description
Double-wall, air-insulated, welded steel construction offers fire resistance in a compact design.
- High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
- Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
- Easy-closing, self-latching doors facilitate fire protection of the cabinet
- Self latch engages critical three-point latching system for protection under fire conditions
- Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
- Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
- SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
- Adjustable leveling feet ensure solid positioning and alignment for tight door closure
- Exterior side panel has built-in grounding connector for easy grounding
- Finished inside and out with durable lead-free powder paint
- 2 in. (5cm) leakproof sump controls spills
- With manual or self-closing door
Order Info
Capacity: 22 gal. (83L). Dimensions: 18L x 23.25W x 65 in.H (46 x 59 x 165cm).
Certifications
FM approved.
Compliance
Meet OSHA and NFPA requirements. Self-closing model complies with Uniform Fire Code.
Warranty
10-Year
Specifications
14.56 in. | |
37 cm | |
18 in. | |
65 in. | |
Slimline storage cabinet | |
Flammables | |
165.1 cm | |
U-Loc™ handle with cylinder lock and key set, 3 Galvanized Steel shelves and 2 in. leakproof sump, dual vents, adjustable leveling feet, a grounding connector and large trilingual warning label | |
1 | |
Epoxy, Polyester | |
90 kg | |
59.05 cm |
45.7 cm | |
Yellow | |
14.56 x 20 x 60.5 in. (37 x 50.8 x 153.7 cm) | |
Steel (CR) | |
23.25 in. | |
60.5 in. | |
153.7 cm | |
22 gal. | |
3 | |
200 lb. | |
20 in. | |
50.8 cm |