Justrite™ Maintenance Spill Berms
To contain minor spills and leaks during temporary storage of drums, machine parts, batteries and equipment
Supplier:  Justrite™ 28410
 View more versions of this product
Catalog No. 19-111-019
Description
Description
  • Made of broad UV and chemical resistant PVC coated fabric
  • No assembly required
  • Berm anchored in place using brass grommets in each corner
  • Encased urethane foam sidewalls for easy roll-through access without need for ramp
  • Walls spring back into place without memory loss
  • Anchor stakes not included
  • Optional grates to keep products and machinery above fluid can be ordered separately
Specifications
Specifications
80 gal.
PVC-coated Fabric
2.4 x 1.2 x 0.1 m
302.8 L
Maintenance Spill Berm