missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Justrite™ Mini-Berm Flex Spill Control Tray

Compact, portable spill control for harsh and hazardous chemicals

$626.04 - $1465.62

Specifications

Material PVC-coated Fabric
Type Spill Control Tray
Includes 1 in. NPT drain fitting/utility strap
View More Specs

Products 4
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Dimensions (L x W x H) Price Quantity  
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Dimensions (L x W x H) Price Quantity  
19-111-029
View Documents
Justrite™
28444
55 gal. 208 L 1.2 x 1.2 x 0.15 m
Each for $969.43
 
19-111-030
View Documents
Justrite™
28446
70 gal. 265 L 1.5 x 1.2 x 0.15 m
Each for $958.60
 
19-111-031
View Documents
Justrite™
28448
110 gal. 416 L 1.8 x 1.5 x 0.15 m
Each for $1,465.62
 
19-111-028
View Documents
Justrite™
28442
30 gal. 113.5 L 1.8 x 1.5 x 0.15 m
Each for $626.04
 
Description

Description

  • For tabletop parts cleaning and fluid transfer operations involving petroleum products and acids
  • Made from robust modified PVC-coated fabric
  • Features rigid corner supports and sturdy foam walls
  • Includes 1 in. NPT drain fitting and utility strap
  • Fluids for disposal can be transfered by attching hose
  • Easy to fold and store

Specifications

Specifications

PVC-coated Fabric
Spill Control Tray
1 in. NPT drain fitting/utility strap
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'