Justrite™ Nonmetallic Dispensing Can
Permit easy, one-hand dispensing of hazardous liquids directly onto work surface. The nonmetallic dispensing can is made from polyethylene with a brass dispenser valve.
$115.85 - $130.33
Specifications
Product Type Dispensing Can
For Use With (Application) For use with Flammables
Material Polyethylene
Includes Leakproof Brass Dispenser Valves Self-close
View More Specs
Products 2
Catalog Number Mfr. No. Color Capacity (English) Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Color Capacity (English) Capacity (Metric) Price Quantity  
17-190B
View Documents
Justrite™
14010
Red 1 qt. 0.95 L
Each for $130.33
17-190A
View Documents
Justrite™
14004
Red 0.5 pt. 0.24 L
Each for $115.85
Description
Description
  • Ideal for use with flammables
  • Leakproof, self-closing dispenser valves minimize spills, controls excess
  • High-density polyethylene body
  • Permits easy, one-hand dispensing of hazardous liquids directly onto work surface
Specifications
Specifications
Dispensing Can
For use with Flammables
Polyethylene
Leakproof Brass Dispenser Valves Self-close
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'