Justrite™ Outdoor Safety Locker Accessory, R-11 Insulations
For use in conjunction with heating and cooling packages to provide optimum temperature control
Supplier:  Justrite™ 915605
 View more versions of this product
Catalog No. 18-666-631
Explore available promotions
Description
Description
  • Installed in ceiling, walls, and sump walls for added temperature control when used in conjunction with air conditioning and heating accessories
  • Must be purchased in conjunction with locker to ensure proper installation
Warranty
One year limited warranty
Specifications
Specifications
R-11 Insulation
Yellow
6-drum, Fire-rated Locker Only