Justrite™ Outdoor Safety Locker Accessory, R-11 Insulations
For use in conjunction with heating and cooling packages to provide optimum temperature control
Supplier: Justrite™ 915607
Description
- Installed in ceiling, walls, and sump walls for added temperature control when used in conjunction with air conditioning and heating accessories
- Must be purchased in conjunction with locker to ensure proper installation
Warranty
One year limited warranty
Specifications
R-11 Insulation | |
Yellow | |
9-drum, Fire-rated Locker Only |