Justrite™ QuickBerm™ Washdown Stations
Portable drive-through station captures contaminated water from vehicle and construction equipment wash down
Supplier: Justrite™ 28578
Description
- Vinyl coated material reinforced with radio frequency-welded, high visibility yellow runway strips that withstand drive-thru traffic, including backhoes and skid steers
- Removable foam inserts of 102mm (4 in.) diameter, supports sidewalls and planks (203W x 51mm H) slide into pockets to keep end walls elevated to prevent wastewater from escaping during drive-over
- Foldable to compact size after removing foam inserts
- Two 1 in. NPT fittings on each side of berm for drainage
- Brass grommets around perimeter enables securely anchor berm during use
Compliance
Comply with EPA for stormwater regulations.
Specifications
Washdown Station | |
425 gal. | |
1609 L | |
5.5 x 3 x 0.1 m |