missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Justrite™ Sure-Grip™ EX Undercounter SS Safety Cabinets for Hazardous Materials
Supplier: Justrite™ 862328
Specifications
Leakproof sills and dual vents | |
Storing hazardous materials | |
35 in. | |
22 in. | |
Blue | |
88.9 cm | |
Self-close | |
2 |
Undercounter | |
35 in. | |
88.9 cm | |
47.6 x 80.6 x 69.5 cm (18.75 x 31.75 x 27.3 in.) | |
55.9 cm | |
22 gal. (83 L) | |
1 | |
NFPA, IFC; FM |