missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kappler™ Zytron™ 200 Hooded Coveralls

Provide increased protection with resistance to a wider range of chemicals

Supplier:  Kappler™ Z2H4143X

 View more versions of this product

Catalog No. 19-171-140E


Explore available promotions

Description

Description

  • Attached hood
  • Zippered front
  • Color: Gray

Applications:

Valuable in applications where rip-out is a concern

Specifications

Specifications

3X-Large
Gray
Elastic
Elastic
Sealed, Taped
ASTM D751, D3787
Ideally used with wider range of chemicals
4.1 kg
Zytron™
Attached Boots
Hooded
Zipper with Double Storm Flap, Hook and Loop Seal
Chemicals
Zipper front, attached hood and boots, elastic wrist and face opening, double storm flap with hook and loop closure
9 lb.
Videos
SDS
Documents

Documents

Promotions

Promotions