missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kappler™ Zytron™ 200 Hooded Coveralls

Provide increased protection with resistance to a wider range of chemicals

Supplier:  Kappler™ Z2B428SMMD

 View more versions of this product

Catalog No. 19-171-142A


Explore available promotions

Description

Description

  • Attached hood
  • Zippered front
  • Color: Gray

Applications:

Valuable in applications where rip-out is a concern

Specifications

Specifications

Medium
Gray
Elastic
Attached
Bound
ASTM D751, D3787
Ideally used with wider range of chemicals
7.3 kg
Zytron™
Elastic
Hooded
Front Zipper
Chemicals
Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening
16 lb.
Videos
SDS
Documents

Documents

Promotions

Promotions