missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kappler™ Zytron™ 200 Hooded Coveralls

Provide increased protection with resistance to a wider range of chemicals

Supplier:  Kappler™ Z2H428LGXLDUP

 View more versions of this product

Catalog No. 19-171-143C


Explore available promotions

Description

Description

  • Attached hood
  • Zippered front
  • Color: Gray

Applications:

Valuable in applications where rip-out is a concern

Specifications

Specifications

X-Large
Gray
Elastic
Attached
Sealed, Taped
ASTM D751, D3787, CE Type 3
Ideally used with wider range of chemicals
4.1 kg
Zytron™
Elastic
Hooded
Front Zipper
Chemicals
Elastic ankle and cuff, single storm flap with pressure sensitive tape closure over the front zipper, attached hood, face opening
9 lb.
Videos
SDS
Documents

Documents

Promotions

Promotions