Dynalon™ Kartell Dry Heat Sterility Indicator Tape
Improve sterilization processes with temperature indicators. The Kartell Dry Heat Sterility Indicator Tape changes color to indicate products have attained temperatures for dry heat sterilization.
Supplier:  Dynalon™ EPD202075
Catalog No. S37250
Explore available promotions
Description
Description
  • For dry heat sterilization
  • After 30 minutes at 155°C, the light green V stamp changes color to brown
  • Indicates sterilization temperature/conditions attained