missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dynalon™ Kartell™ Pipet/Buret Washing System Parts
Parts for Kartell™ Automatic Pipet Washing Set.
Supplier: Dynalon™ 215905
Requires : Tubing not included with washer.
Description
Two or three baskets can be used with one or two jars and one rinser for economy of operation. Since the soaking operation takes the longest, two or three batches of glassware can be soaking at the same time.
- Hold pipets or burets up to 24” (60.8cm) long
- Polyethylene construction reduces breakage and buffers the tops of pipets against the bottom of jars
- Back-siphoning is eliminated because water inlet is located at top of rinser
- Diameter of tubing permits water to drain completely
- Rinser repeats automatically
For complete unit, see S31880A. S31880 is washer unit only and requires S318801 and S318802. Tubing not included.
Order Info
Dimensions: 6.5 dia. x 5.5 in. H (16.5 x 65cm).
Specifications
25.5 in. | |
6.5 in. | |
Polyethylene | |
Pipet or Buret Washer |
64.77 cm | |
16.5cm | |
For Kartell™ automatic pipet washing set |