missing translation for 'onlineSavingsMsg'
Learn More

KCl Electrolyte Solution, METTLER TOLEDO™

Maintenance and pH electrolyte solution

Supplier:  METTLER TOLEDO 51350072

Numéro de catalogue. 01912583


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • 3M KCl
  • pH electrolyte and maintenance solution
  • 250 mL
Spécifications

Spécifications

Electrolyte, KCl
7447-40-7
KCl
74.55
7
Colorless
770°C
3 M
Plastic Bottle
250 mL
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions