KD Scientific Stainless-Steel Syringes
Durable, electron-beam construction withstands high force, high pressure applications
Supplier:  KD Scientific 780801
 View more versions of this product
Catalog No. 14-931-35
Explore available promotions
Description
Description
Eliminates hazards of glass syringe breakage.
  • Compatible with most syringe pumps
  • Fully autoclavable for reuse
  • Adaptable to Luer Lock or Swagelok™ fittings
  • Resistant to most chemicals
Specifications
Specifications
Autoclavable
19/100 in.
12.7 mm
Syringe Pump
Stainless Steel (316)
7000 psi
2.5 mL
1/2 in.
4.8 mm
None
Swagelok 1/16 in. Syringe