KD Scientific Stainless-Steel Syringes
Durable, electron-beam construction withstands high force, high pressure applications
Supplier:  KD Scientific 780816
 View more versions of this product
Catalog No. 14-931-41
Explore available promotions
Description
Description
Eliminates hazards of glass syringe breakage.
  • Compatible with most syringe pumps
  • Fully autoclavable for reuse
  • Adaptable to Luer Lock or Swagelok™ fittings
  • Resistant to most chemicals
Specifications
Specifications
Autoclavable
3/4 in.
22.3 mm
Syringe Pump
Stainless Steel (316)
1500 psi
20 mL
22/25 in.
19 mm
None
Luer-Lok Syringe
Product Suggestions
Product Suggestions