missing translation for 'onlineSavingsMsg'
Learn More

Kemtec™ Building Simple DC Circuits with Real Electronic Components Kit

Teach simple DC circuitry using real components and help students relate projects to real world applications.

Supplier:  Customer Channels Group (Ccg) 15224

Catalog No. S07353


Only null left
Explore available promotions

Alert:

Choking hazard (1) Small parts. Not for children under 3 yrs.


Includes: Red LED, multimeter, solderless breadboard, 9V battery, test lead wire, 9V battery snap, 330 Ohm resistor, 820 Ohm resistor, 560 Ohm resistor, 680 Ohm resistor, 220 Ohm (1/2 watt) resistor, 150 Ohm resistor, matches, instructor manual

Requires : Impact goggles

Description

Description

This basic kit introduces the use of easy-to-manipulate, reusable, solder-less breadboards, resistors, LEDs, jump wires and batteries, allowing hands-on experience with circuitry components for individual students.

  • Students learn to design circuits and read circuit diagrams
  • Students can investigate effects of resistors and LEDs in real circuits
  • Instructor's manual included with reproducible student guides covering stepwise experimental protocols, simple circuit diagrams, homework assignments and assessment materials
  • Includes separate assignments for middle and high school levels
  • Complete unit with five experiments, lecture information, sample data, and complete answer keys
  • Complete kit also includes one digital multimeter for teacher to demonstrate how to measure voltage, current and resistance so that data may be collected for problem
  • Available as single kit (1 to 4 students) or class-pack kit (24 students)
  • Grade 7 to 10

Specifications

Specifications

7 to 10
Teach electronic circuitry using real components and help students relate projects to real life applications
5 experiments, lecture information, sample data, complete answer keys, real electronic components kit, instructor's manual
Class-pack Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions