missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Keratinocyte SFM (1X)
Contains L-glutamine.
Supplier: Gibco™ 17005042
Description
Keratinocyte-SFM is supplied with prequalified human recombinant Epidermal Growth Factor 1-53 (EGF 1-53) and Bovine Pituitary Extract (BPE) in separate packaging. Components are not sold separately.
Note:
A Keratinocyte-SFM Master File has been submitted to the FDA. Permission to cross-reference the Master File may be obtained by contacting Technical Services or your local Sales Representative.
Specifications
• 500 mL Keratinocyte-SFM Basal Medium; store in refrigerator (2â8°C) and protect from light • 25 mg Bovine Pituitary Extract (BPE); store in freezer (-5 to -20°C) and protect from light • 2.5 μg EGF, Human Recombinant; store in freezer (-5 to -20°C) and protect from light Complete Medium: store in refrigerator (2â8°C) and protect from light |
|
Serum-free | |
Mammalian Cell Culture | |
Kit | |
Sterile-filtered | |
Glutamine, Phenol Red |
Keratinocyte Cells, Epithelial Cells | |
1 X | |
Liquid | |
Serum-free | |
Sterile-filtered | |
500 mL |
For Research Use Only. Not for use in diagnostic procedures.