missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α-Ketoglutaric acid sodium salt, 98%, Thermo Scientific Chemicals

$121.40 - $121.40

Chemical Identifiers

CAS 22202-68-2
Molecular Formula C5H5NaO5
Molecular Weight (g/mol) 168.08
InChI Key MOTOGHHLNTXPTI-UHFFFAOYSA-M
Synonym sodium hydrogen 2-oxoglutarate, pentanedioic acid, 2-oxo-, sodium salt, pentanedioic acid, 2-oxo-, sodium salt 1:?, 2-ketoglutaric acid monosodium salt, 2-oxoglutaric acid, sodium salt, glutaric acid, 2-oxo-, sodium salt, alpha-ketoglutaric acid, sodium salt, 2-oxoglutaric acid 1-sodium salt, sodium 4-carboxy-4-oxobutanoate
PubChem CID 23672314
IUPAC Name sodium;5-hydroxy-4,5-dioxopentanoate
SMILES C(CC(=O)[O-])C(=O)C(=O)O.[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC439350050
SDS View Documents
Thermo Scientific Chemicals
439350050
5 g
Each for $121.40
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

22202-68-2
168.08
sodium hydrogen 2-oxoglutarate, pentanedioic acid, 2-oxo-, sodium salt, pentanedioic acid, 2-oxo-, sodium salt 1:?, 2-ketoglutaric acid monosodium salt, 2-oxoglutaric acid, sodium salt, glutaric acid, 2-oxo-, sodium salt, alpha-ketoglutaric acid, sodium salt, 2-oxoglutaric acid 1-sodium salt, sodium 4-carboxy-4-oxobutanoate
sodium;5-hydroxy-4,5-dioxopentanoate
C5H5NaO5
MOTOGHHLNTXPTI-UHFFFAOYSA-M
23672314
C(CC(=O)[O-])C(=O)C(=O)O.[Na+]

Specifications

22202-68-2
97.5
3.6 to 4.0 (0.5% aq. soln.)
5 g
C5H5NaO5
sodium hydrogen 2-oxoglutarate, pentanedioic acid, 2-oxo-, sodium salt, pentanedioic acid, 2-oxo-, sodium salt 1:?, 2-ketoglutaric acid monosodium salt, 2-oxoglutaric acid, sodium salt, glutaric acid, 2-oxo-, sodium salt, alpha-ketoglutaric acid, sodium salt, 2-oxoglutaric acid 1-sodium salt, sodium 4-carboxy-4-oxobutanoate
C(CC(=O)[O-])C(=O)C(=O)O.[Na+]
168.08
168.08
Authentic
Glass Bottle
White
100.0
Powder
98%
HO2CCH2CH2C(=O)CO2Na
MOTOGHHLNTXPTI-UHFFFAOYSA-M
sodium;5-hydroxy-4,5-dioxopentanoate
23672314
98%
1.0% max.
α-Ketoglutaric acid sodium salt
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 244-836-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only