missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ketoprofen 98.0+%, TCI America™
$306.92 - $306.92
Chemical Identifiers
CAS | 22071-15-4 |
---|---|
Molecular Formula | C16H14O3 |
Molecular Weight (g/mol) | 254.285 |
MDL Number | MFCD00055790 |
InChI Key | DKYWVDODHFEZIM-UHFFFAOYSA-N |
Synonym | ketoprofen, 2-3-benzoylphenyl propanoic acid, orudis, m-benzoylhydratropic acid, 2-3-benzoylphenyl propionic acid, ketoprofene, profenid, oruvail, 3-benzoylhydratropic acid, alrheumat |
PubChem CID | 3825 |
ChEBI | CHEBI:6128 |
IUPAC Name | 2-(3-benzoylphenyl)propanoic acid |
SMILES | CC(C1=CC=CC(=C1)C(=O)C2=CC=CC=C2)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
K003825G
|
TCI America
K003825G |
25 g |
N/A
|
|
|||||
Chemical Identifiers
22071-15-4 | |
254.285 | |
DKYWVDODHFEZIM-UHFFFAOYSA-N | |
3825 | |
2-(3-benzoylphenyl)propanoic acid |
C16H14O3 | |
MFCD00055790 | |
ketoprofen, 2-3-benzoylphenyl propanoic acid, orudis, m-benzoylhydratropic acid, 2-3-benzoylphenyl propionic acid, ketoprofene, profenid, oruvail, 3-benzoylhydratropic acid, alrheumat | |
CHEBI:6128 | |
CC(C1=CC=CC(=C1)C(=O)C2=CC=CC=C2)C(=O)O |
Specifications
22071-15-4 | |
White | |
MFCD00055790 | |
2811 | |
DKYWVDODHFEZIM-UHFFFAOYSA-N | |
2-(3-benzoylphenyl)propanoic acid | |
3825 | |
254.29 | |
Crystalline Powder |
95°C | |
C16H14O3 | |
25 g | |
ketoprofen, 2-3-benzoylphenyl propanoic acid, orudis, m-benzoylhydratropic acid, 2-3-benzoylphenyl propionic acid, ketoprofene, profenid, oruvail, 3-benzoylhydratropic acid, alrheumat | |
CC(C1=CC=CC(=C1)C(=O)C2=CC=CC=C2)C(=O)O | |
254.285 | |
CHEBI:6128 | |
≥98.0% (HPLC,T) | |
Ketoprofen |
Safety and Handling
RTECSNumber : UE7570000
TSCA : Yes