missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Keyed Alike Safety Lock with Plastic Shackle
Help ensure workers stay safe from hazardous energy during servicing and repair with strong, efficient and cost-effective Safety Lockout Padlocks from Brady.
Supplier: Brady™ SDPLRED38PLKA12
Description
- Designed exclusively for lockout tagout applications as part of an OSHA-compliant safety program.
- Key-retaining brass cylinder prevents key removal in the unlocked position for added safety and helps limit lost keys.
- Nylon padlocks with plastic shackles are lightweight, non-conductive, non-sparking and ideal for electrical or general lockout applications.
- Locks that are Keyed Alike (KA) reduce the number of keys needed to carry and are ideal for individuals or trades responsible for multiple machines or isolation points.
- Record and retain engraved key codes to streamline replacement of lost keys.
Specifications
Padlock | |
Red | |
0.25 in. | |
0.82 x 1.5 x 1.77 in. | |
(12) Padlocks, (12) Keys, (12) English Labels, (12) French Labels, (12) Spanish Labels | |
6 |
Keyed Alike | |
Nylon | |
6.35 mm | |
38.10 mm | |
Plastic | |
12 Pk. |