missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Keyed Alike Safety Lock with Steel Shackle
Help ensure workers stay safe from hazardous energy during servicing and repair with strong, efficient and cost-effective Safety Lockout Padlocks from Brady.
Supplier: Brady™ SDPLGRN38STKA6
Description
- Designed exclusively for lockout tagout applications as part of an OSHA-compliant safety program.
- Key-retaining brass cylinder prevents key removal in the unlocked position for added safety and helps limit lost keys.
- Nylon padlocks with steel shackles are lightweight, durable and ideal for general lockout applications.
- Locks that are Keyed Alike (KA) reduce the number of keys needed to carry and are ideal for individuals or trades responsible for multiple machines or isolation points.
- Record and retain engraved key codes to streamline replacement of lost keys.
Specifications
Padlock | |
Green | |
0.25 in. | |
0.82 x 1.5 x 1.77 in. | |
38.10 mm | |
Steel | |
6 Pk. |
Keyed Alike | |
Nylon | |
6.35 mm | |
Ideal for electrical contractors, maintenance, electricians | |
(6) Padlocks, (6) Keys, (6) English Labels, (6) French Labels, (6) Spanish Labels | |
6 |