missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Keystone Safety KeyGuard Microporous Lab Coats
Latex free microporous lab coats are a cost savings alternative to Tyvek and provide contamination control in a variety of environments including cleanrooms.
Supplier: Keystone Safety LCOWEKG4XL
Description
- Non-linting
- Microporous
- Latex free
- Cost savings alternative to Tyvek brand
- Individually inspected for stitching, sizing, material and workmanship
- Ideal for cleanroom environments including electronic, medical device, and pharmaceutical industries where garment control is a concern
Specifications
Controlled Environments Lab Coat | |
Unisex | |
White | |
Elastic | |
Non-sterile | |
Yes | |
Serged | |
30/Pk. |
4X-Large | |
Microporous Film | |
Snaps | |
Single-use | |
None | |
LCO-WE-KG | |
Manufacturing, Food Service, Industrial, Medical, Pharmaceutical | |
Keyguard |