missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ki-67 Recombinant Rat Monoclonal Antibody (SolA15), Alexa Fluor™ Plus 555, Invitrogen™
Rat Recombinant Monoclonal Antibody
Supplier: Fisher Chemical 740008TP555
Description
Ki-67 Recombinant Monoclonal antibody specifically detects Ki-67 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ Plus recombinant antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning data. Alexa Fluor™ Plus antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor™ Plus antibodies provide brighter signal compared to leading Alexa Fluor™ antibodies, providing you with better signal-to-noise for your critical experiments. These antibodies show better specificity and lot-to-lot consistency as these are recombinant antibodies, generated by cloning specific genes for the desired antibodies into an expression vector and expressed in vitro. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Applications Tested: This SolA15 antibody has been tested by immunohistochemistry of mouse spleen tissue and immunocytochemistry and flow cytometric analysis of HeLa cells. This may be used at 0.02-0.25 μg per test for flow cytometry. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 553 nm; Emission: 568 nm; Laser: Yellow Laser Filtration: 0.2 μm post-manufacturing filtered.Specifications
Ki-67 | |
Recombinant Monoclonal | |
1.0 mg/mL | |
proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.8 | |
E9PVX6, P46013 | |
Rat | |
50 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
SolA15 | |
Alexa Fluor Plus 555 | |
Mki67 | |
antigen identified by monoclonal antibody Ki 67; antigen identified by monoclonal antibody Ki-67; Antigen identified by monoclonal antibody Ki-67 homolog; Antigen KI-67; Antigen KI-67 homolog; antigen KI-67; proliferation marker protein Ki-67; antigen KI-67-like; cb31; D630048A14Rik; I79_022666; Ki67; Ki-67; KIA; LOW QUALITY PROTEIN: proliferation marker protein Ki-67; marker of proliferation Ki-67; MIB-; MIB-1; Mki67; PPP1R105; Proliferation marker protein Ki-67; proliferation-related Ki-67 antigen; protein phosphatase 1, regulatory subunit 105; RP11-380J17.2; sb:cb31; si:ch211-250b22.7; unnamed protein product; wu:fa11g09; wu:fb57a07; wu:fi14e05 | |
Protein G | |
RUO | |
17345, 4288 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |