missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Ki-67 Recombinant Rat Monoclonal Antibody (SolA15), Biotin, Invitrogen™
Rat Recombinant Monoclonal Antibody
$350.10 - $960.35
Spécifications
Antigen | Ki-67 |
---|---|
Clone | SolA15 |
Concentration | 1.0 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Recombinant Monoclonal |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
74-000-8TB
|
Fisher Chemical
740008TB |
50 μL |
chaque for $960.35
|
|
|||||
50-264-8623
|
Fisher Chemical
740008TB20UG |
20 μL |
chaque for $350.10
|
|
|||||
Description
Ki-67 Recombinant Monoclonal antibody specifically detects Ki-67 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Recombinant Rat monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Spécifications
Ki-67 | |
1.0 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.4 | |
antigen identified by monoclonal antibody Ki 67; antigen identified by monoclonal antibody Ki-67; Antigen identified by monoclonal antibody Ki-67 homolog; Antigen KI-67; Antigen KI-67 homolog; antigen KI-67; proliferation marker protein Ki-67; antigen KI-67-like; cb31; D630048A14Rik; I79_022666; Ki67; Ki-67; KIA; LOW QUALITY PROTEIN: proliferation marker protein Ki-67; marker of proliferation Ki-67; MIB-; MIB-1; Mki67; PPP1R105; Proliferation marker protein Ki-67; proliferation-related Ki-67 antigen; protein phosphatase 1, regulatory subunit 105; RP11-380J17.2; sb:cb31; si:ch211-250b22.7; unnamed protein product; wu:fa11g09; wu:fb57a07; wu:fi14e05 | |
IgG2a κ | |
Protein G | |
Antibody |
SolA15 | |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Biotin | |
Rat | |
Human, Mouse | |
E9PVX6, P46013 | |
17345, 4288 | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Mki67 |