DWK Life Sciences Kimble™ Kim-Kap™ Closures
Available in natural (clear) or choice of four colors
Supplier:  DWK Life Sciences 7366213
 View more versions of this product
Catalog No. 14-957-92
Explore available promotions
Description
Description
Available in natural (clear) or choice of four colors
  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable
Specifications
Specifications
Push Cap
Polypropylene
Non-sterile
Reusable
Autoclavable
Red
13 mm O.D. Plain-end Culture Tubes