missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ KIMAX™ Acid-Test Burets

Shows readings directly in percentage of acidity

Supplier:  DWK Life Sciences A620F1

Catalog No. 11-301-40


Explore available promotions

Description

Description

  • PTFE stopcock plug
  • Graduated from 0 to 1%
  • Capacity: 10mL

Specifications

Specifications

10 mL
0.1 mL
PTFE Stopcock Plug, No. 2 Stopcock
±0.05 mL
ASTM E542, ASTM E694, NIST
Borosilicate Glass
Volumetric Analysis
PTFE
Acid Automatic Buret
Videos
SDS
Documents

Documents

Promotions

Promotions