missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Kimble™ KIMAX™ Griffin Beakers

With thick, beaded, slightly flared top

Supplier:  DWK Life Sciences 14000800

 View more versions of this product

Catalog No. 02-539N



Explore available promotions

Description

Description

With thick, beaded, slightly flared top

  • With pouring spout
  • Matte-finish enameled marking spot
  • 20 to 4000mL sizes have white scales for approximate volume
  • Accurate to ±5%
  • Uniform, sturdy walls

Specifications

Specifications

±5%
800 mL
For Measuring and Mixing Liquids
Spout
Matte Finish Enamel Marking
Beaker
ASTM E960, Type I
Griffin Beaded Top
27.05 oz.
Borosilicate Glass
50 to 750 mL
50 mL
6 Pk.
Yes
Dual White Scale
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions