Kimble™ Plastic Test Tubes
An economical alternative to glass.
Supplier:  Kimble™ 51012200E
 View more versions of this product
Catalog No. S32168
Explore available promotions
Description
Description
Plastic test tubes can be used in place of glass for most routine laboratory procedures.
Polypropylene tubes
  • Can be centrifuged up to 3000rcf
  • Autoclavable up to 120°C for 15 minutes
  • Tolerate most common acids and solvents at room temperature
Polystyrene tubes
  • Not autoclavable
  • Can be centrifuged up to 1400rcf
  • Tolerate aqueous solutions, mild bases and weak acids
Order Info
Sold in packs of 250.
Specifications
Specifications
Test Tube
3000 x G
75 mm
Non-sterile
12 mm
Autoclavable
Polypropylene