DWK Life Sciences Kimble™ KIMAX™ Reusable Test Tubes with Beaded Rim
Made from N-51A borosilicate glass
Supplier:  DWK Life Sciences 4505025150
 View more versions of this product
Catalog No. 14-915G
Explore available promotions
Description
Description
  • Uniform wall thickness for maximum heat transfer and chemical resistance
Specifications
Specifications
ASTM E438, Type I, Class B
55 mL
Borosilicate Glass
25 mm
Plain Beaded Rim
150 mm
None