DWK Life Sciences Kimble™ KIMAX™ Reusable Test Tubes with Beaded Rim
Made from N-51A borosilicate glass
Supplier: DWK Life Sciences 4505025200
Description
- Uniform wall thickness for maximum heat transfer and chemical resistance
Specifications
ASTM E438, Type I, Class B | |
75 mL | |
Borosilicate Glass | |
25 mm |
Plain Beaded Rim | |
200 mm | |
None |