Learn More
Kimble™ KIMAX™ Test Tubes with Lip and Marking Spot
Durable Kimax borosilicate tubes.
$269.00 - $269.00
Specifications
Certifications/Compliance ASTM E982, Type I
Design Type Plain
Material Borosilicate Glass
Label Type Marking Spot
View More Specs
Products 1
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Capacity (Metric) Price Quantity  
S30614A
View Documents
Kimble™
4504220150
20 mm 150 mm 36 mL
Pack of 72 Each for $269.00
Case of 8 Pack for $2,152.00
Only null left
Description
Description
Quality glass test tubes with lip and ceramic enamel marking spot.
Specifications
Specifications
ASTM E982, Type I
Plain
Borosilicate Glass
Marking Spot
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'