Kimberly-Clark Professional™ G60 Level 5 Cut-Resistant Sleeves
Kimberly-Clark™ Professional G60 Level 5 Cut-Resistant Sleeves are for industrial tasks that require protection against arm lacerations
Supplier:  Kimberly-Clark Professional™ 90075
Catalog No. 19-028-322
Explore available promotions
Description
Description
  • Offer the highest level of cut protection in a sleeve: EN388 Level 5 Cut Resistance / ANSI Cut Level 3 , with CPPT = 1,244  grams
  • Coated; Made of ultra-high molecular weight polyethylene (UHMWPE) fiber, which enhances fit and breathability
  • Proprietary closure on the bicep and wrist provides an adjustable and custom fit (one size fits most )
  • Typical uses include metal fabrication, glass handling and automotive assembly
Specifications
Specifications
Dyneema Fiber, Lycra Fiber, Glass, Nylon
ANSI Cut Level 3 with CPPT of 1244 grams
18 in.
EN388 Cut Level 5
Closure
45.7 cm