Learn More
KleenGuard™ G60 Medium Duty Polyurethane-Coated Cut-Resistant Gloves
KleenGuard™ G60 Medium Duty Polyurethane-Coated Cut-Resistant Gloves are constructed of ultra-high molecular weight polyethylene fibers to enhance breathability, and provide ANSI Cut Level 3 protection.
Supplier:  KleenGuard™ 13824
 View more versions of this product
Catalog No. 18-042-923
Only null left
Explore available promotions
Description
Description
  • Constructed of ultra-high molecular weight polyethylene (UHMWPE) fibers to provide breathability and protection
  • Polyurethane coated palm and fingers enhance dexterity and grip capabilities
  • Separate right and left gloves in a pair provide a snug fit
Specifications
Specifications
ANSI 4
Gray
Specific
HPPE
Medium
Assembly, plastics, light raw materials
13824
13
24 cm
Polyurethane
ANSI A2
Yes
Cut-resistant Gloves
EN 388
Knit
KleenGuard™ G60
9.5 in.
Product Suggestions
Product Suggestions