missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KleenGuard™ G60 Medium Duty Polyurethane-Coated Cut-Resistant Gloves
KleenGuard™ G60 Medium Duty Polyurethane-Coated Cut-Resistant Gloves are constructed of ultra-high molecular weight polyethylene fibers to enhance breathability, and provide ANSI Cut Level 3 protection.
Supplier: KleenGuard™ 13827
Description
- Constructed of ultra-high molecular weight polyethylene (UHMWPE) fibers to provide breathability and protection
- Polyurethane coated palm and fingers enhance dexterity and grip capabilities
- Separate right and left gloves in a pair provide a snug fit
Specifications
| ANSI 4 | |
| Gray | |
| Specific | |
| HPPE | |
| 2X-Large | |
| Assembly, plastics, light raw materials | |
| 13827 | |
| 13 | |
| 27 cm |
| Polyurethane | |
| ANSI A2 | |
| Yes | |
| Cut-resistant Gloves | |
| EN 388 | |
| Knit | |
| KleenGuard™ G60 | |
| 10.6 in. |