Kimberly-Clark Professional™ KleenGuard™ G60 Polyurethane Coated Cut-Resistant Gloves, ANSI Level A2
Black and white polyurethane coated gloves are ideal for industrial tasks that require protection against minor hand lacerations and abrasions.
Supplier:  Kimberly-Clark Professional™ 38692
 View more versions of this product
Catalog No. 19-005-453
Explore available promotions
Description
Description
  • Offer EN388 Level 3 / ANSI Level 2 Cut Resistance and CPPT = 686 grams
  • Made of ultra-high molecular weight polyethylene (UHMWPE) yarn with nylon and Lycra fiber, which provides both cut resistance and a comfortable fit
  • Polyurethane coating enhances grip and the dark color helps hide dirt for longer wear
  • Each pair has a dedicated right and left glove
Specifications
Specifications
Polyurethane
ANSI A2
Cut-resistant Gloves
EN 388 Cut Level 3
Knit
G60
24.4 cm
Black, White
Dyneema, Fiber, Lycra
X-Large
Metal fabrication, Glass handling, aerospace and automotive assembly
38692
9.6 in.