missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark Professional™ KC500 Kimguard™ Sterilization Wraps

Offers precisely right protection for specific needs

$731.20 - $731.20

Specifications

Color Dark Blue
For Use With (Application) Used to package linen towels, packs, gowns, light basin sets and single instruments for sterilization
Material SMS Fabric
Sterility Non-sterile
Latex Free Yes
View More Specs

Products 1
Catalog Number Mfr. No. Length (English) Width (English) Price Quantity  
Catalog Number Mfr. No. Length (English) Width (English) Price Quantity  
19-168-207
View Documents
Kimberly-Clark Professional™
68118
18 in. 18 in.
Case of 600 Each for $731.20
 
Description

Description

Offers precisely right protection for specific needs

  • Sequential wrap
  • Used for packaging of linen towels, packs, gowns, light basin sets, and single instruments for sterilization
  • Low-lint fabric enables prevention from contamination after sterilization
  • Steam and EO compatible
  • Naturally latex-free
  • Non-sterile
  • Available in multiple sizes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.

Order Info

Case count indicates the number of sheets included.

Specifications

Specifications

Dark Blue
SMS Fabric
Yes
Used to package linen towels, packs, gowns, light basin sets and single instruments for sterilization
Non-sterile
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'