missing translation for 'onlineSavingsMsg'
Learn More

Ansell Kimtech Pure™ A5 Sterile Cleanroom Apparel

100% polypropylene breathable SMS fabric has cloth-like feel

$683.98 - $873.94

Specifications

Color White
Closure Type Snap
Seam Type Bound
Description Tunnelized elastic, built-in thumb loop, folded inside out pattern or cuffed
For Use With (Application) ISO Class 5 or higher cleanroom environment
View More Specs

Products 9
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-057-475
Accessories View Documents
Ansell
88800
Small
Case of 25 Each for $683.98
Only null left
 
19-057-476
Accessories View Documents
Ansell
88801
Medium
Case of 25 Each for $683.98
Only null left
 
19-057-490
Accessories View Documents
Ansell
88802
Large
Case of 25 Each for $715.36
Only null left
 
19-057-491
Accessories missing translation for 'viewDocuments'
Ansell
88803
X-Large
Case of 25 Each for $715.36
Only null left
 
19-057-492
Accessories missing translation for 'viewDocuments'
Ansell
88804
2X-Large
Case of 25 Each for $693.41
Only null left
 
19-057-493
Accessories missing translation for 'viewDocuments'
Ansell
88805
3X-Large
Case of 25 Each for $768.23
Only null left
 
19-057-494
Accessories missing translation for 'viewDocuments'
Ansell
88806
4X-Large
Case of 25 Each for $798.90
Only null left
 
18-042-910
missing translation for 'viewDocuments'
Ansell
12914
6X-Large
Case of 25 Each for $873.94
Only null left
 
18-042-911
missing translation for 'viewDocuments'
Ansell
12917
5X-Large
Case of 25 Each for $865.38
Only null left
 
Description

Description

100% polypropylene breathable SMS fabric has cloth-like feel

Kimtech™ A5 Sterile Cleanroom Coveralls have an average bacterial filtration efficiency (BFE) of 96% and are made of 100% polypropylene breathable SMS fabric, which gives the wearer cloth-like comfort.

The Reflex Coverall Design is 7.5 times more rip-resistant than ANSI minimum standards and offers excellent fit and freedom of movement for the wearer. This personal protective apparel is triple-bagged and vacuum packaged, suitable for ISO Class 5 or higher cleanrooms.

It is not made with natural rubber latex, silicone, or the BHT preservative and is designed for single-use (disposable PPE). The easy-to-don, individually packaged coveralls feature a mandarin collar, thumb loops and tunneled elastics.

  • 25 Kimtech™ A5 Sterile Cleanroom Coveralls/Case
  • Triple bagged, vacuum packed, suitable for ISO Class 5 or higher cleanrooms; CE certified (Category III)
  • Reflex Coverall Design offers excellent fit and freedom of movement and 7.5 times more rip-resistant than ANSI minimums
  • This protective apparel isn't made with natural rubber latex, BHT preservative or silicone
  • Kimtech™ A5 Cleanroom Apparel is manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions
Specifications

Specifications

White
Bound
ISO Class 5 or higher cleanroom environment
Snap
Tunnelized elastic, built-in thumb loop, folded inside out pattern or cuffed
Videos
SDS
Documents

Documents