Kimberly-Clark Professional™ KleenGuard™ A10 Coveralls
A10 Coveralls provide economical, disposable protection for workers exposed to dirt and grime in the workplace.
Supplier:  Kimberly-Clark Professional™ 12230
 View more versions of this product
Catalog No. 19-041-845
Explore available promotions
Description
Description
  • Designed for entry-level protection from hazardous dust
  • Feature a zipper front, attached hood, serged seams and elastic wrists
  • Five layers of stacked barrier protection, for a lightweight, breathable protective coverall
  • Ideal for food processing, general maintenance and manufacturing
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Hooded
Serged
ANSI ISEA 101-1996
Food Processing, General Maintenance, Manufacturing
4X-Large
Polypropylene
Single-use
Elastic
Front Zipper
Non-sterile
Yes