Ansell KleenGuard™ A20 Breathable Particle Protection Frocks
Protection against dry particulates and light liquid sprays
Supplier:  Ansell 36265
 View more versions of this product
Catalog No. 19-150-1615
Explore available promotions
Description
Description
  • Long, abrasion-resistant jackets made of breathable SMMMS fabric - 5 layers of stacked barrier protection
  • Raglan sleeves with elastic wrists and high collar
  • No pockets
  • Plastic snap front closure
  • White
Specifications
Specifications
2X-Large
White
Suited for laboratories, decontamination modesty garment, remediation, pesticides, heavy dust environments
46 in.
SMS
Snaps
NFPA 99
117cm