missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark Professional™ KleenGuard™ A20 Frocks

Frocks feature breathable, patented MICROFORCE barrier SMS fabric and meets NFPA 99 criteria for antistatic materials.

$343.36 - $343.36

Specifications

Product Type General Purpose Frock
Material SMS
Color White
Cuff Style Elastic
For Use With (Application) Laboratories, Decontamination, Remediation, Pesticides, Heavy Dust Environments
View More Specs

Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-457-221
View Documents
Kimberly-Clark Professional™
34815
5X-Large
Case of 30 Each for $343.36
 
Description

Description

  • Snap front closure with snap at collar
  • Long length
  • Elastic wrists
  • No pockets
  • Passes NFPA 99 criteria for antistatic materials
  • Typical uses: Laboratories; Decontamination modesty garment; Remediation; Pesticides; Heavy dust environments

Order Info

This size is available on a made-to-order basis – 30 case minimum order per product code; product available within 8 weeks of order receipt

Specifications

Specifications

General Purpose Frock
White
Laboratories, Decontamination, Remediation, Pesticides, Heavy Dust Environments
SMS
Elastic
NFPA 99
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'