missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark Professional™ Electronic Cassette Skin Care Dispenser

For use with Kleenex™ brand skin cleansers and hand sanitizers

$218.69 - $218.69

Specifications

Color Black
View More Specs

Products 1
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
19-413-103
View Documents
Kimberly-Clark Professional™
11430
Floor Stand Dispenser
Case of 2 Each for $218.69
 
Description

Description

  • Reliable touchless operation reduces cross contamination and the spread of germs
  • Long battery life dispenses 60,000 single uses
  • Reliable controlled dispensing reduces waste
  • Key or push button lock
  • Made of durable ABS plastic

Specifications

Specifications

Black
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'