Kimberly-Clark Professional™ Jackson Safety™ Cap Liners
Thermal cap liners protect against wind and chill in environments ranging from moderate to bitter cold.
Supplier:  Kimberly-Clark Professional™ 16760
 View more versions of this product
Catalog No. 29-636-2157
Explore available promotions
Description
Description
Universal fit. Hook-and-loop chin closures and two suspension fastener straps.
Lightweight Liner
  • 100% cotton twill shell
  • Heavyweight fleece lining for additional warmth
Heavyweight Liner
  • Detachable flame resistant neck piece made of two layers of spun polyester with two-button neck closure
  • 100% cotton twill shell
  • Flame retardant headpiece lined with red fleece
  • Fits over hearing protectors even when used with four-point suspension hats and caps
  • Dielectric
Heavyweight flame-retardant liner retains flame-retardant quality after washing.
Specifications
Specifications
Fleece-lined Cotton Twill