Kimberly-Clark Professional™ KleenGuard™ V80 MonoGoggle™ XTR OTG Safety Goggles
Comfort and splash protection are at your fingertips with the KleenGuard V80 Monogoggle XTR OTG Goggles. The safety splash goggles give you eye protection from liquids with a sporty design and curved lens for an enhanced vision field.
Supplier:  Kimberly-Clark Professional™ 18624
Catalog No. 29-636-2114
Explore available promotions
Description
Description
  • Stylish goggle protects against splashes, while providing increased visibility due to its cylindrical lens
  • Soft, pliable frame conforms to the face for comfort and is easy to clean
  • Indirect ventilation
  • Can be worn over most prescription eyeglasses
  • Clear polycarbonate lens with antifog coating
  • Blue frame
Specifications
Specifications
Meets ANSI Z87.1+ standards for high impact (18624-02 meets ANSI Z87.1+2010 and 18624-03 meets ANSI Z87.1+2015) with a D3 rating for droplet and splash protection
Safety Goggles; XTR Goggle
Indirect
Clear
Blue
Polycarbonate