Ansell Kimtech™ A5 Non-Sterile Integrated Hood and Mask
Hood and mask combination guards against the risk of contamination in cleanroom environments.
Supplier:  Ansell 49828
Catalog No. 19-166-702
Explore available promotions
Description
Description
  • Integrated hood and mask sonically welded together combines two donning steps into one
  • Stretch fit elastic headband for more secure fit
  • Pull-down ties in back to assist in the gowning process
  • Latex- and rubber-free
  • Universal size
  • White
  • Bulk packed
Specifications
Specifications
Universal
White
Single-use
49828
Bulk
Pull down ties
Class 100 (ISO 5)
Polypropylene SMS Fabric
Ties
Yes
Non-sterile
75/Cs.
Eyes Only
Product Suggestions
Product Suggestions