Kimberly-Clark Professional™ KleenGuard™ A20 Breathable Particle Protection Lab Coats
A20 lab coats are comprised of five layers of stacked barrier protection (SMMMS) against hazardous particles. They are a popular choice for dusty environments such as laboratories and remediation, as well as pesticide applications and as decontamination modesty garments.
Supplier:  Kimberly-Clark Professional™ 35623
 View more versions of this product
Catalog No. 19-457-326
Explore available promotions
Description
Description
A20 lab coats are comprised of five layers of stacked barrier protection (SMMMS) against hazardous particles. They are a popular choice for dusty environments such as laboratories and remediation, as well as pesticide applications and as decontamination modesty garments.
  • Tailored style made of breathable patented MICROFORCE™ Barrier SMS Fabric
  • Knee-length, serged seams
  • Long sleeves, notched-lapel collar
  • Air and moisture vapor can pass through to aid wearer comfort
  • Meets NFPA 99 criteria for antistatic materials
  • Two styles to choose from:
    • Open wrists with pockets, snap closure
    • Elastic wrists without pockets, hook and loop closure
Compliance
NFPA 99 compliant antistatic material
Specifications
Specifications
General Purpose Lab Coat
Unisex
White
Elastic
Non-sterile
None
35623
NFPA 99
1.6 oz. Fabric
3X-Large
SMS
Hook and Loop
Limited-use
Knee-length
Notched Lapel
Serged
Laboratories, Remediation, Decontamination, Pesticides